Merck
  • Organocatalytic asymmetric epoxidation and tandem epoxidation/Passerini reaction under eco-friendly reaction conditions.
Organocatalytic asymmetric epoxidation and tandem epoxidation/Passerini reaction under eco-friendly reaction conditions.
Organic & biomolecular chemistry (2012-08-25)
Anna Maria Deobald, Arlene G Corrêa, Daniel G Rivera, Márcio Weber Paixão
ABSTRACT
An eco-friendly synthesis of highly functionalized epoxides and their incorporation into an organocatalytic multicomponent approach are reported. For this, a modified class of diarylprolinol silyl ethers was designed to enable high catalytic activity in an environmentally benign solvent system. The one-pot procedure showed great efficiency in promoting stereoselective multicomponent transformations in a tandem, 'green' fashion. Because of its non-residual, efficient and selective character, this synthetic design shows promise for large-scale applications in both diversity and target-oriented syntheses.
Sigma-Aldrich
(S)-(+)-2-Pyrrolidinemethanol, 97%