Merck
  • Aldol reactions catalyzed by L-proline functionalized polymeric nanoreactors in water.
Aldol reactions catalyzed by L-proline functionalized polymeric nanoreactors in water.
Chemical communications (Cambridge, England) (2012-08-23)
Annhelen Lu, Pepa Cotanda, Joseph P Patterson, Deborah A Longbottom, Rachel K O'Reilly
ABSTRACT
The use of functional core-shell micelles as asymmetric catalytic nanoreactors for organic reactions in water is presented. An unprecedented increase in rate of reaction was achieved, which is proposed to be associated with the ability of the nanostructures to effectively concentrate the reagents in the catalytically active micelle core.
Product Description
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, 99.8%
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)