Merck
  • Pore surface engineering with controlled loadings of functional groups via click chemistry in highly stable metal-organic frameworks.
Pore surface engineering with controlled loadings of functional groups via click chemistry in highly stable metal-organic frameworks.
Journal of the American Chemical Society (2012-08-22)
Hai-Long Jiang, Dawei Feng, Tian-Fu Liu, Jian-Rong Li, Hong-Cai Zhou
ABSTRACT
Reactions of ZrCl(4) and single or mixed linear dicarboxylic acids bearing methyl or azide groups lead to highly stable isoreticular metal-organic frameworks (MOFs) with content-tunable, accessible, reactive azide groups inside the large pores. These Zr-based MOFs offer an ideal platform for pore surface engineering by anchoring various functional groups with controlled loadings onto the pore walls via the click reaction, endowing the MOFs with tailor-made interfaces. Significantly, the framework and crystallinity of the functionalized MOFs are well-retained, and the engineered pore surfaces have been demonstrated to be readily accessible, thus providing more opportunities for powerful and broad applications of MOFs.
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%
Sigma-Aldrich
Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%