Merck
  • Infrared to visible upconversion in Ho³+/Yb³+ co-doped Y₂ O₃ phosphor: effect of laser input power and external temperature.
Infrared to visible upconversion in Ho³+/Yb³+ co-doped Y₂ O₃ phosphor: effect of laser input power and external temperature.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-21)
Monika Rai, K Mishra, S K Singh, R K Verma, S B Rai
ABSTRACT
In the present paper, Ho(3+) doped and Ho(3+)/Yb(3+) co-doped Y(2)O(3) phosphors have been synthesized using solution combustion technique and characterized for its structure and upconversion (UC) fluorescence as a function of Yb(3+) concentration. Effect of a variation in laser input power and external temperature on the UC emission intensity has been studied to explore the UC mechanism and temperature dependent behavior of the phosphor, respectively. On excitation with near infrared (NIR) light (976 nm), the phosphor emits strong green emission along with relatively weak emission bands in red and blue regions at 553, 670 and 497 nm due to (5)S(2)→(5)I(8), (5)F(5)→(5)I(8) and (5)F(3)→(5)I(8), respectively. The emission shows a decrease in intensity with an increase in external temperature, however contrary to the normal behavior of Ho(3+), no significant change in the FIR (fluorescence intensity ratio) of (5)F(4)→(5)I(8) and (5)S(2)→(5)I(8) transitions is noted in the present host. This peculiar behavior of the sample with external temperature has been explained by temperature dependent lifetime study of the thermally coupled levels.
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, 99.999% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, nanopowder, <50 nm particle size
Sigma-Aldrich
Yttrium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Yttrium(III) oxide, dispersion, 10 wt. % in isopropanol, nanoparticles, <100 nm (DLS), ≥99.9% trace metals basis