Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Insecticidal activity of five commercial mosquito coils against Anopheles albimanus, Aedes albopictus, and Culex quinquefasciatus.
Insecticidal activity of five commercial mosquito coils against Anopheles albimanus, Aedes albopictus, and Culex quinquefasciatus.
Journal of the American Mosquito Control Association (2012-08-17)
Rui-De Xue, Whitney A Qualls, Jessica D Phillips, Tong-Yan Zhao
ABSTRACT
The insecticidal activity of 4 mosquito coils containing pyrethroid ester insecticides and 1 mosquito coil containing 15% citronella oil was evaluated against adult female Anopheles albimanus, Aedes albopictus, and Culex quinquefasciatus. Two of the pyrethroid mosquito coils containing 0.08% meperfluthrin showed the strongest insecticidal activity, and all 4 pyrethroid coils produced significantly higher mortality than the citronella coil across all 3 test species.