Merck
  • Synthesis of 2-Aryl benzothiazoles via K2S2O8-mediated oxidative condensation of benzothiazoles with aryl aldehydes.
Synthesis of 2-Aryl benzothiazoles via K2S2O8-mediated oxidative condensation of benzothiazoles with aryl aldehydes.
The Journal of organic chemistry (2012-07-28)
Zhiyong Yang, Xiang Chen, Sizhuo Wang, Jidan Liu, Kai Xie, Anwei Wang, Ze Tan
ABSTRACT
Nontransition metal-catalyzed synthesis of 2-aryl benzothiazoles was achieved through K(2)S(2)O(8)-mediated oxidative condensation of benzothiazoles with aryl aldehydes. The same transformation can also be effected when the aryl aldehydes were replaced with phenylglyoxylic acids.
Sigma-Aldrich
Phenylglyoxylic acid, 97%
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Phenylglyoxylic acid, purum, ≥98.0% (T)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%