Merck
  • The effects of chronic ingestion of mercuric chloride on fertility and testosterone levels in male Sprague Dawley rats.
The effects of chronic ingestion of mercuric chloride on fertility and testosterone levels in male Sprague Dawley rats.
Journal of biomedicine & biotechnology (2012-07-26)
John C Heath, Y Abdelmageed, Tim D Braden, Hari O Goyal
ABSTRACT
Although male infertility is well researched, the effects of inorganic mercury on male reproduction and fertility are less well known. Studies pertaining to mercury and male fertility identified reduced concentration of testosterone in the serum of male workers, a toxic influence on fertility of organic mercury compounds within concentrations at the workplace, and increased days to pregnancy. We evaluated the effect of chronic mercuric chloride (HgCl(2)) exposure in male rats on reproductive endpoints. Thirty-day old male Sprague Dawley rats (n = 31) were exposed to 0.0, 1.0, or 2.0 mg/kg/day of HgCl(2) via gavage. After 60 days exposure, they were housed with nonexposed females for 21 days. A survivor analysis revealed the exposed animals took longer to impregnate the females and had a lower rate of impregnation. Further statistical analysis revealed a lower correlation between testicular testosterone levels and days to impregnate, and also lower sperm counts in the epididymis head and body of the exposed males. The results indicate that HgCl(2) exposure had significant adverse effects on male rat reproduction endpoints including fertility at a dose that was not clinically toxic.
Sigma-Aldrich
Mercury(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
Mercury(II) chloride, ACS reagent, ≥99.5%