Merck
  • A Zn2+ specific triazole based calix[4]arene conjugate (L) as a fluorescence sensor for histidine and cysteine in HEPES buffer milieu.
A Zn2+ specific triazole based calix[4]arene conjugate (L) as a fluorescence sensor for histidine and cysteine in HEPES buffer milieu.
The Analyst (2012-07-24)
Rakesh Kumar Pathak, Khatija Tabbasum, Ankit Rai, Dulal Panda, Chebrolu Pulla Rao
ABSTRACT
A highly fluorescent Zn(2+) complex of the triazole linked salicyl-imino-thiophenyl conjugate of calix[4]arene, [ZnL] has been demonstrated to be a chemo-sensing ensemble for the recognition of His and Cys among the naturally occurring amino acids in HEPES buffer milieu. The recognition behaviour of the [ZnL] towards these amino acids has been shown on the basis of fluorescence, absorption and visual fluorescent colour changes. The species of recognition were shown by ESI MS titrations, AFM & TEM microscopy and cell studies.
Product Description
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
HEPES sodium salt, Vetec, reagent grade, 96%
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture