Merck
  • Analysis of initial reactions of MALDI based on chemical properties of matrixes and excitation condition.
Analysis of initial reactions of MALDI based on chemical properties of matrixes and excitation condition.
The journal of physical chemistry. B (2012-07-18)
Yin-Hung Lai, Chia-Chen Wang, Chiu Wen Chen, Bo-Hong Liu, Sheng Hsien Lin, Yuan Tseh Lee, Yi-Sheng Wang
ABSTRACT
This investigation concerns the initial chemical reactions that affect the ionization of matrixes in matrix-assisted laser desorption/ionization (MALDI). The study focuses on the relaxations of photon energy that occur on a comparable time scale to that of ionization, in which the available laser energy is shared and the ionization condition is changed. The relaxations include fluorescence, fragmentation, and nonradiative relaxation from the excited state to the ground state. With high absorption cross section and long excited-state lifetime, photoionization of matrix plays an important role if sufficient laser energy is used. Under other conditions, thermal ionization of the molecule in the ground state is predicted to be one of the important reactions. Evidence of change in the branching ratio of initial reactions with the matrix and the excitation wavelength was obtained with α-cyano-4-hydroxycinnamic acid, sinapinic acid, 2,5-dihydroxybenzoic acid, and 2,4,6-trihydroxyacetophenone. These matrixes are studied by obtaining their mixed crystal absorption spectra, fluorescence properties, laser-induced infrared emission, and product ions. The exact ionization pathway depends on the chemical properties of matrixes and the excitation conditions. This concept may explain the diversity of experimental results observed in MALDI experiments, which provides an insight into the ensemble of chemical reactions that govern the generation of ions.
Product Description
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
Gentisic acid sodium salt hydrate, ≥98%
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
trans-Cinnamic acid, analytical standard
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%