Merck
  • Rapid determination of thiamine, riboflavin, niacinamide, pantothenic acid, pyridoxine, folic acid and ascorbic acid in Vitamins with Minerals Tablets by high-performance liquid chromatography with diode array detector.
Rapid determination of thiamine, riboflavin, niacinamide, pantothenic acid, pyridoxine, folic acid and ascorbic acid in Vitamins with Minerals Tablets by high-performance liquid chromatography with diode array detector.
Journal of pharmaceutical and biomedical analysis (2012-07-13)
Pengfei Jin, Lufeng Xia, Zheng Li, Ning Che, Ding Zou, Xin Hu
ABSTRACT
A simple, isocratic, and stability-indicating high-performance liquid chromatography (HPLC) method has been developed for the rapid determination of thiamine (VB(1)), niacinamide (VB(3)), pyridoxine (VB(6)), ascorbic acid (VC), pantothenic acid (VB(5)), riboflavin (VB(2)) and folic acid (VB(9)) in Vitamins with Minerals Tablets (VMT). An Alltima C(18) column (250 mm × 4.6 mm i.d., 5 μm) was used for the separation at ambient temperature, with 50mM ammonium dihydrogen phosphate (adjusting with phosphoric acid to pH 3.0) and acetonitrile as the mobile phase at the flow rate of 0.5 ml min(-1). VB(1), VB(3), VB(6), VC and VB(5) were extracted with a solution containing 0.05% phosphoric acid (v/v) and 0.3% sodium thiosulfate (w/v), and were then simultaneously analyzed by using the mobile phase of phosphate buffer-acetonitrile (95:5, v/v), while VB(2) and VB(9) were extracted with a solution containing 0.5% ammonium hydroxide solution (v/v), and were then simultaneously analyzed by using the mobile phase of phosphate buffer-acetonitrile (85:15, v/v). The detection wavelengths were 275 nm for VB(1), VB(3), VB(6), VC, 210 nm for VB(5), and 282 nm for VB(2) and VB(9). The method showed good system suitability, sensitivity, linearity, specificity, precision, stability and accuracy. All the seven water-soluble vitamins were well separated from other ingredients and degradation products. Method comparison indicated good concordance between the developed method and the USP method. The developed method was reliable and convenient for the rapid determination of VB(1), VB(3), VB(6), VC, VB(5), VB(2) and VB(9) in VMT.
Sigma-Aldrich
Sodium thiosulfate pentahydrate, 99.999% trace metals basis
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium thiosulfate pentahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Ammonium hydroxide solution, puriss., meets analytical specification of Ph. Eur., 25-30% NH3 basis
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a., reag. ISO, reag. Ph. Eur., ~25% NH3 basis
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonium hydroxide solution, puriss. p.a. plus, ≥25% NH3 in H2O