Merck
  • Interaction of LED light with coinitiator-containing composite resins: effect of dual peaks.
Interaction of LED light with coinitiator-containing composite resins: effect of dual peaks.
Journal of dentistry (2012-07-10)
Jae-Seong Sim, Hyo-Joung Seol, Jeong-Kil Park, Franklin Garcia-Godoy, Hyung-Il Kim, Yong Hoon Kwon
ABSTRACT
Recently the colour stability of composite resins has been an issue due to the emphasis on the aesthetics of restored teeth. The purpose of the present study was to investigate how dual-peak LED units affect the polymerization of coinitiator-containing composite resins. Five composite resins [coinitiator-containing: Aelite LS Posterior (AL), Tetric EvoCeram (TE), and Vit-l-escence (VI); only CQ-containing: Grandio (GD) and Filtek Z350 (Z3)] were light cured using four different light-curing units (LCUs). Among them, Bluephase G2 (BP) and G-light (GL) were dual-peak LED LCUs. Microhardness, polymerization shrinkage, flexural, and compressive properties were measured. BP and GL had no consistent effect on the microhardness of AL, TE, and VI on the top and bottom surfaces of resin specimens. Among the specimens, AL and VI showed the least (9.86-10.41 μm) and greatest (17.58-19.21 μm) polymerization shrinkage, respectively. However, the effect of BP and GL on the shrinkage of specimens was not consistent. Among the specimens, GD showed the greatest flexural properties [strength (FS) and modulus (FM)] and TE showed the lowest flexural and compressive properties [strength (CS) and modulus (CM)]. In same resin product, maximum FS and CS differences due to the different LCUs were 10.3-21.0% and 3.6-9.2%, respectively. Furthermore, the influences of BP and GL on FS and CS were not consistent. The tested dual-peak LED LCUs had no consistent synergic effect on the polymerization of coinitiator-containing composite resins as compared with QTH and single-peak LED LCUs. The dual-peak LED LCUs achieve a similar degree of polymerization in coinitiator-composite resins as QTH and single-peak LED LCUs did. Choice of LCU does not appear to be a determinant of the light curing of coinitiator-composite resins.
Product Description
Supelco
D-Camphor, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(1S)-(−)-Camphor, 95%
Sigma-Aldrich
D-Camphor, ≥97%, FG
Sigma-Aldrich
(1S)-(+)-Camphorquinone, 99%
Supelco
(−)-Camphor, analytical standard
Sigma-Aldrich
(±)-Camphor, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Sigma-Aldrich
(±)-Camphor, ≥95.5%
Sigma-Aldrich
Camphor, 96%
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 98%, FG
Sigma-Aldrich
1-Phenyl-1,2-propanedione, 99%
Sigma-Aldrich
(1R)-(+)-Camphor, 98%
Camphor (dl), primary reference standard
Camphor (racemic), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Camphor, purum, synthetic, ≥95.0% (GC)