Merck
  • FeCl2-promoted cleavage of the unactivated C-C bond of alkylarenes and polystyrene: direct synthesis of arylamines.
FeCl2-promoted cleavage of the unactivated C-C bond of alkylarenes and polystyrene: direct synthesis of arylamines.
Angewandte Chemie (International ed. in English) (2012-06-13)
Chong Qin, Tao Shen, Conghui Tang, Ning Jiao
ABSTRACT
Ironing it out: an efficient and convenient nitrogenation strategy involving C-C bond cleavage for the straightforward synthesis of versatile arylamines is presented. Various alkyl azides and alkylarenes, including the common industrial by-product cumene, react using this protocol. Moreover, this method provides a potential strategy for the degradation of polystyrene.
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)