Merck
  • Impact of carbon source on nitrous oxide emission from anoxic/oxic biological nitrogen removal process and identification of its emission sources.
Impact of carbon source on nitrous oxide emission from anoxic/oxic biological nitrogen removal process and identification of its emission sources.
Environmental science and pollution research international (2012-06-12)
Zhen Hu, Jian Zhang, Shanping Li, Huijun Xie
ABSTRACT
Wastewater treatment is an important source of nitrous oxide (N(2)O), which is a strong greenhouse gas and dominate ozone-depleting substance. The purpose of this study was to evaluate the effect of carbon source on N(2)O emission from anoxic/oxic biological nitrogen removal process. The mechanisms of N(2)O emission were also studied. Long-term experiments were operated to evaluate the effect of three different carbon sources (i.e., glucose, sodium acetate, and soluble starch) on N(2)O emission characteristics. And batch experiments, in the presence or absence of specific inhibitors, were carried out to identify the sources of N(2)O emission. The ammonia-oxidizing bacteria (AOB) and denitrifiers community compositions under different circumstances were also analyzed based on which the underlying mechanisms of N(2)O emission were elucidated. The conversion ratios of N(2)O in reactors with glucose, sodium acetate, and soluble starch were 5.3 %, 8.8 %, and 2.8 %, respectively. The primary process responsible for N(2)O emission was nitrifier denitrification by Nitrosomonas-like AOB, while denitrification by heterotrophic denitrifiers acted as the sink. Reactor with sodium acetate showed the highest N(2)O emission, together with the highest nitrogen and phosphate removal ratios. Carbon source has a significant impact on N(2)O emission quantity and relatively minor effect on its production mechanism.
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, Vetec, reagent grade, 98%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%