Merck
  • High-contrast photopatterning of photoluminescence within quantum dot films through degradation of a charge-transfer quencher.
High-contrast photopatterning of photoluminescence within quantum dot films through degradation of a charge-transfer quencher.
Advanced materials (Deerfield Beach, Fla.) (2012-06-09)
Mario Tagliazucchi, Victor A Amin, Severin T Schneebeli, J Fraser Stoddart, Emily A Weiss
ABSTRACT
Diffraction-limited, high-contrast photopatterning of the photoluminescence of layer-by-layer films comprising CdSe@CdS@ZnS quantum dots and polyviologen is reported. The photoluminescence of the quantum dots is initially quantitatively quenched due to ultrafast photoinduced electron transfer to polyviologen. Photopatterning is achieved by high-power or prolonged illumination in air, which photochemically degrades the polyviologen and thereby restores the photoluminescence of the quantum dots.
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O