Merck
  • Ordered mesoporous silica material SBA-15: loading of new calcium channel blocker--lacidipine.
Ordered mesoporous silica material SBA-15: loading of new calcium channel blocker--lacidipine.
Journal of microencapsulation (2012-06-09)
A Kiwilsza, J Mielcarek, A Pajzderska, J Wąsicki
ABSTRACT
Mesoporous material SBA-15 of hexagonal structure was synthesised and its usefulness as a carrier for a poorly soluble drug--lacidipine (LA)--was tested. The source of silica was tetraethyl orthosilicate (TEOS) and the structure ordering agent was non-ionic surfactant Pluronic P123. SBA-15 with encapsulated LA was characterised by X-ray diffraction (XRD), infrared spectroscopy (FTIR), thermogravimetry (TG), differential scanning calorimetry (DSC) and transmission electron microscopy (TEM). Adsorption of the therapeutically active substance was performed in anhydrous environment (chloroform). The content of the therapeutic substance in SBA-15 estimated from TG measurements reached 9%. The properties of the compound obtained were compared with those of a physical mixture of the components.
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Poloxamer 188
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Lacidipine, ≥98% (HPLC)