Merck
  • Identification of gene clusters associated with fusaric acid, fusarin, and perithecial pigment production in Fusarium verticillioides.
Identification of gene clusters associated with fusaric acid, fusarin, and perithecial pigment production in Fusarium verticillioides.
Fungal genetics and biology : FG & B (2012-06-02)
Daren W Brown, Robert A E Butchko, Mark Busman, Robert H Proctor
ABSTRACT
The genus Fusarium is of concern to agricultural production and food/feed safety because of its ability to cause crop disease and to produce mycotoxins. Understanding the genetic basis for production of mycotoxins and other secondary metabolites (SMs) has the potential to limit crop disease and mycotoxin contamination. In fungi, SM biosynthetic genes are typically located adjacent to one another in clusters of co-expressed genes. Such clusters typically include a core gene, responsible for synthesis of an initial chemical, and several genes responsible for chemical modifications, transport, and/or regulation. Fusarium verticillioides is one of the most common pathogens of maize and produces a variety of SMs of concern. Here, we employed whole genome expression analysis and utilized existing knowledge of polyketide synthase (PKS) genes, a common cluster core gene, to identify three novel clusters of co-expressed genes in F. verticillioides. Functional analysis of the PKS genes linked the clusters to production of three known Fusarium SMs, a violet pigment in sexual fruiting bodies (perithecia) and the mycotoxins fusarin C and fusaric acid. The results indicate that microarray analysis of RNA derived from culture conditions that induce differential gene expression can be an effective tool for identifying SM biosynthetic gene clusters.
Sigma-Aldrich
Fusaric acid, from Gibberella fujikuroi
Pricing and availability is not currently available.