Merck
  • Development and bioactivity evaluation of bioglasses with low Na2O content based on the system Na 2O-CaO-MgO-P 2O 5-SiO 2.
Development and bioactivity evaluation of bioglasses with low Na2O content based on the system Na 2O-CaO-MgO-P 2O 5-SiO 2.
Journal of materials science. Materials in medicine (2012-06-01)
Emad El-Meliegy, Esmat M A Hamzawy, Abeer M El-Kady, Aida Salama, Ahalam El-Rashedi
ABSTRACT
Osteoconductive bioglasses, free of K(2)O and Al(2)O(3) and with content of Na(2)O lower than 10 mol%, were designed based on the ratio (SiO(2) + MgO)/(P(2)O(5) + CaO + Na(2)O) in the system Na(2)O-CaO-MgO-P(2)O(5)-SiO(2). The developed glasses have shown a strong potential for the formation of hydroxycarbonated apatite (HCA) in vitro. The particles of HCA aggregates tend to be of finer size with increasing the ratio of (SiO(2) + MgO)/(CaO + P(2)O(5) + Na(2)O) in the glass chemical composition indicating significant bioactivity. Critical size bone defects created in the femurs of albino adult female rats, and grafted with the glass particles for 12 weeks post implantation, were completely healed by filling with mineralized bone matrix without infection showing a strong potential for new bone formation in vivo. Osteoblasts and osteocytes were observed close to the surface of the granular implants with active areas of bone deposition, resorption and remodelling. The bioglass with lowest (SiO(2) + MgO)/(CaO + P(2)O(5) + Na(2)O) ratio has shown the highest bioactivity while the bioglass with the highest (SiO(2) + MgO)/(CaO + P(2)O(5) + Na(2)O) has shown the lowest bioactivity. The newly formed bone in vivo has shown a similar structure to that of the original bone as indicated by the histology and microstructural results. In addition, Ca/P molar ratio of the newly formed bone was found to be (~1.67), which is similar to that of the original bone.
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Sodium oxide, 80%