Merck
  • Deep Raman spectroscopy for the non-invasive standoff detection of concealed chemical threat agents.
Deep Raman spectroscopy for the non-invasive standoff detection of concealed chemical threat agents.
Talanta (2012-05-23)
Emad L Izake, Biju Cletus, William Olds, Shankaran Sundarajoo, Peter M Fredericks, Esa Jaatinen
ABSTRACT
Deep Raman spectroscopy has been utilized for the standoff detection of concealed chemical threat agents from a distance of 15 m under real life background illumination conditions. By using combined time and space resolved measurements, various explosive precursors hidden in opaque plastic containers were identified non-invasively. Our results confirm that combined time and space resolved Raman spectroscopy leads to higher selectivity towards the sub-layer over the surface layer as well as enhanced rejection of fluorescence from the container surface when compared to standoff spatially offset Raman spectroscopy. Raman spectra that have minimal interference from the packaging material and good signal-to-noise ratio were acquired within 5 s of measurement time. A new combined time and space resolved Raman spectrometer has been designed with nanosecond laser excitation and gated detection, making it of lower cost and complexity than picosecond-based laboratory systems.
Sigma-Aldrich
Nitromethane, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Nitromethane, ACS reagent, ≥95%
Sigma-Aldrich
Nitromethane, suitable for HPLC, ≥96%
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium nitrate, BioXtra, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, suitable for plant cell culture
Sigma-Aldrich
Ammonium nitrate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium nitrate, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium nitrate, ≥99.0%