Merck
  • Determination of monensin in milk samples by front-surface long-wavelength fluoroimmunoassay using nile blue-doped silica nanoparticles as labels.
Determination of monensin in milk samples by front-surface long-wavelength fluoroimmunoassay using nile blue-doped silica nanoparticles as labels.
Talanta (2012-05-23)
Juan Godoy-Navajas, Maria Paz Aguilar-Caballos, Agustina Gómez-Hens
ABSTRACT
A heterogeneous immunoassay for monensin determination in milk samples using a tracer formed by anti-monensin antibodies bound to nile blue (NB)-doped silica nanoparticles (NPs), 96-well microplates as solid supports and long-wavelength fluorescence measurements is described for the first time. The assay relies on the competition of the monensin present in the samples with a monensin-bovine serum albumin conjugate, which was immobilized onto the well surface, for the active sites of anti-monensin antibodies. After subsequent incubation and washing steps, the fluorescence of the bound tracer fraction is measured onto the dry surface of the well. An antigen capture format was also assayed by immobilizing anti-sheep IgG previously to the incubation of sheep anti-monensin antibodies and using a tracer formed by monensin bound to nile blue-doped silica NPs, which competes with the analyte for binding the immobilized antibody. Although the fluorescence signal obtained in both formats can be correlated to the analyte concentration, better results were obtained using the antibody capture format. After the optimization of the system using this format, the method features a detection limit of 0.015 μg L(-1) and a dynamic range from 0.05 to 5 μg L(-1). The precision, assayed at two different analyte concentrations, 0.2 and 1 μg L(-1), and expressed as relative standard deviation, gave values of 5.9% and 4.0%, respectively. The method was satisfactorily applied to the analysis of milk samples, which only required a simple extraction step in order to remove the proteins from samples, giving recoveries in the range 83.3-107.5%.
Sigma-Aldrich
Monensin sodium salt, 90-95% (TLC)
Sigma-Aldrich
Nile Blue A, certified by the Biological Stain Commission
Sigma-Aldrich
Nile Blue A, Dye content ≥75 %
Sigma-Aldrich
Nile Blue A perchlorate, Dye content 95 %
Sigma-Aldrich
Nile Blue chloride, Dye content 85 %
Supelco
Monensin sodium salt hydrate, VETRANAL®, analytical standard