Merck
  • Rapid purification and characterization of γ-glutamyl-transpeptidase from shiitake mushroom (Lentinus edodes).
Rapid purification and characterization of γ-glutamyl-transpeptidase from shiitake mushroom (Lentinus edodes).
Journal of food science (2012-05-18)
Jianrong Li, Ju Huang, Jie Yin, Ning Wu, Jun Song, Lei Zhang, Tianjia Jiang
ABSTRACT
γ-Glutamyl-transpeptidase (GGT) is one of the important enzymes in the pathway of odor formation in shiitake mushroom (Lentinus edodes). Rapid purification and characterization of GGT from shiitake mushroom were studied in this work. The GGT was purified 179-fold after 3 primary steps: precipitation by ammonium sulfate, isolation by Phenyl Sepharose 6 FF, and desalting by Sephadex G-25. The enzyme, consisting of a small and a large subunit with Mr 28 KDa and 60 KDa, respectively, is composed of 17 kinds of amino acids with the ratio of basic and acidic residues 1: 1.84, and its secondary structure was also determined by Fourier transform infrared spectroscopy. The properties of GGT were studied with γ-glutamyl-p-nitroanilide as the substrate. The results showed Km value of 2.601μM, optimal temperature of 40 °C, and isoelectric point of 6.4. In addition, the activity of GGT was promoted by Na⁺, K⁺, and Ca²⁺ and inhibited by Cu²⁺, Ag⁺, Zn²⁺, and Fe³⁺. In this work, the γ-glutamyl-transpeptidase (GGT) from shiitake mushroom was purified with a simple scheme. Through the characterization of GGT, the relationship between endogenous formaldehyde and odor formation is to be clarified and assist in finding means of formaldehyde control in shiitake mushroom.
Sigma-Aldrich
Sephadex® G-25, Coarse
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-25, Fine
Sigma-Aldrich
Sephadex® G-75, Medium
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
L-Glutamic acid γ-(p-nitroanilide) hydrochloride, γ-glutamyl transpeptidase substrate
Sigma-Aldrich
L-Glutamic acid γ-(4-nitroanilide), γ-glutamyl transpeptidase substrate
Sigma-Aldrich
Sephadex® G-25, BioReagent, for molecular biology, DNA grade, Superfine
Sigma-Aldrich
Sephadex® G-100, BioReagent, for molecular biology, DNA grade, medium
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-100, Superfine
Sigma-Aldrich
Sephadex® G-50, Fine
Sigma-Aldrich
Sephadex® G-10, Medium
Sigma-Aldrich
Sephadex® G-50, Superfine
Sigma-Aldrich
Sephadex® G-50, BioReagent, for molecular biology, DNA grade, fine
Sigma-Aldrich
Sephadex® G-50, BioReagent, for molecular biology, DNA grade, medium