Merck
  • Development of a fast screening and confirmatory method by liquid chromatography-quadrupole-time-of-flight mass spectrometry for glucuronide-conjugated methyltestosterone metabolite in tilapia.
Development of a fast screening and confirmatory method by liquid chromatography-quadrupole-time-of-flight mass spectrometry for glucuronide-conjugated methyltestosterone metabolite in tilapia.
Journal of agricultural and food chemistry (2012-05-03)
Kande Amarasinghe, Pak-Sin Chu, Eric Evans, Renate Reimschuessel, Nicholas Hasbrouck, Hiranthi Jayasuriya
ABSTRACT
This paper describes the development of a fast method to screen and confirm methyltestosterone 17-O-glucuronide (MT-glu) in tilapia bile. The method consists of solid-phase extraction (SPE) followed by high-performance liquid chromatography-mass spectrometry. The system used was an Agilent 6530 Q-TOF with an Agilent Jet stream electrospray ionization interface. The glucuronide detected in the bile was characterized as MT-glu by comparison with a chemically synthesized standard. MT-glu was detected in bile for up to 7 days after dosing. Semiquantification was done with matrix-matched calibration curves, because MT-glu showed signal suppression due to matrix effects. This method provides a suitable tool to monitor the illegal use of methyltestosterone in tilapia culture.
Sigma-Aldrich
17α-Methyltestosterone, solid (photosensitive)
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Sigma-Aldrich
17α-Methyltestosterone, ≥97.0% (HPLC)