Merck
  • Benzylamine antihyperglycemic effect is abolished by AOC3 gene invalidation in mice but not rescued by semicarbazide-sensitive amine oxidase expression under the control of aP2 promoter.
Benzylamine antihyperglycemic effect is abolished by AOC3 gene invalidation in mice but not rescued by semicarbazide-sensitive amine oxidase expression under the control of aP2 promoter.
Journal of physiology and biochemistry (2012-05-02)
Sandra Grès, Sandy Bour, Philippe Valet, Christian Carpéné
ABSTRACT
Semicarbazide-sensitive amine oxidase (SSAO) is a transmembrane enzyme that metabolizes primary amines from endogenous or dietary origin. SSAO is highly expressed in adipose, smooth muscle and endothelial cells. In each of these cell types, SSAO is implicated in different biological functions, such as glucose transport activation, extracellular matrix maturation and leucocyte extravasation, respectively. However, the physiological functions of SSAO and their involvement in pathogenesis remain uncompletely characterized. To better understand the role of adipose tissue SSAO, we investigated whether it was necessary and/or sufficient to produce the antihyperglycemic effect of the SSAO-substrate benzylamine, already reported in mice. Therefore, we crossed SSAO-deficient mice invalidated for AOC3 gene and transgenic mice expected to express human SSAO in an adipocyte-specific manner, under the control of aP2 promoter. The aP2-human AOC3 construct (aP2-hAOC3) was equally expressed in the adipose tissue of mice expressing or not the native murine form and almost absent in other tissues. However, the corresponding SSAO activity found in adipose tissue represented only 20 % that of control mice. As a consequence, the benzylamine antihyperglycemic effect observed during glucose tolerance test in control was abolished in AOC3-KO mice but not rescued in mice expressing aP2-hAOC3. The capacity of benzylamine or methylamine to activate glucose uptake in adipocytes exhibited parallel variations in the corresponding genotypes. Although the aP2-hAOC3 construct did not allow a total rescue of SSAO activity in adipose tissue, it could be assessed from our observations that adipocyte SSAO plays a pivotal role in the increased glucose tolerance promoted by pharmacological doses of benzylamine.
Sigma-Aldrich
Benzylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Methylamine solution, 2.0 M in methanol
Supelco
Benzylamine, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
Benzylamine hydrochloride
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Benzylamine, ReagentPlus®, 99%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine-13C hydrochloride, 99 atom % 13C
Sigma-Aldrich
Methylamine, ≥99.0%
Sigma-Aldrich
Methylamine-15N hydrochloride, 98 atom % 15N
Sigma-Aldrich
Methylamine hydrochloride, ≥98%