Merck
  • Complete genome sequence of the 2,3-butanediol-producing Klebsiella pneumoniae strain KCTC 2242.
Complete genome sequence of the 2,3-butanediol-producing Klebsiella pneumoniae strain KCTC 2242.
Journal of bacteriology (2012-04-27)
Sang Heum Shin, Sewhan Kim, Jae Young Kim, Soojin Lee, Youngsoon Um, Min-Kyu Oh, Young-Rok Kim, Jinwon Lee, Kap-Seok Yang
ABSTRACT
Here we report the full genome sequence of Klebsiella pneumoniae KCTC 2242,consisting of a 5.26-Mb chromosome (57.6% GC%; 5,035 genes [4,923 encoding known proteins, 112 RNA genes]) and a 202-kb plasmid (50.2% GC%; 229 genes [229 encoding known proteins]).
Sigma-Aldrich
2,3-Butanediol, 98%
Sigma-Aldrich
(2S,3S)-(+)-2,3-Butanediol, 97%
Sigma-Aldrich
(2R,3R)-(−)-2,3-Butanediol, 97%
Sigma-Aldrich
meso-2,3-Butanediol, 99%