Merck
  • Effects of acepromazine maleate on platelet function assessed by use of adenosine diphosphate activated- and arachidonic acid- activated modified thromboelastography in healthy dogs.
Effects of acepromazine maleate on platelet function assessed by use of adenosine diphosphate activated- and arachidonic acid- activated modified thromboelastography in healthy dogs.
American journal of veterinary research (2012-04-27)
Bobbi J Conner, Rita M Hanel, Bernard D Hansen, Alison A Motsinger-Reif, Makoto Asakawa, Clifford R Swanson
ABSTRACT
To evaluate the effect of acepromazine maleate administered IV on platelet function assessed in healthy dogs by use of a modified thromboelastography assay. 6 healthy adult mixed-breed dogs. Dogs received each of 3 treatments (saline [0.9% NaCl] solution [1 to 2 mL, IV] and acepromazine maleate [0.05 and 0.1 mg/kg, IV]) in a randomized crossover study with a minimum 3-day washout period between treatments. From each dog, blood samples were collected via jugular venipuncture immediately before and 30 and 240 minutes after administration of each treatment. A modified thromboelastography assay, consisting of citrated kaolin-activated (baseline assessment), reptilase-ADP-activated (ADP-activated), and reptilase-arachidonic acid (AA)-activated (AA-activated) thromboelastography, was performed for each sample. Platelet inhibition was evaluated by assessing the percentage change in maximum amplitude for ADP-activated or AA-activated samples, compared with baseline values. Percentage change in maximum amplitude was analyzed by use of Skillings-Mack tests with significance accepted at a family-wise error rate of P < 0.05 by use of Bonferroni corrections for multiple comparisons. No significant differences were found in the percentage change of maximum amplitude from baseline for ADP-activated or AA-activated samples among treatments at any time. Platelet function in dogs, as assessed by use of a modified thromboelastography assay, was not inhibited by acepromazine at doses of 0.05 or 0.1 mg/kg, IV. This was in contrast to previous reports in which it was suggested that acepromazine may alter platelet function via inhibition of ADP and AA.
Sigma-Aldrich
Acepromazine maleate, ≥98% (HPLC)
Pricing and availability is not currently available.