Merck
  • Asymmetric total synthesis and absolute stereochemistry of the neuroactive marine macrolide palmyrolide A.
Asymmetric total synthesis and absolute stereochemistry of the neuroactive marine macrolide palmyrolide A.
Organic letters (2012-04-06)
Rodolfo Tello-Aburto, Emily M Johnson, Cheyenne K Valdez, William A Maio
ABSTRACT
The first asymmetric total synthesis and determination of the absolute configuration for the neuroactive marine macrolide palmyrolide A is described. The highlight of the synthesis is macrocyclization via trans-enamide formation catalyzed by copper(I) iodide and cesium carbonate. Comparison with the authentic spectral data confirms the synthesis of (+)-ent-palmyrolide A.
Product Description
Copper(I) iodide ChemBeads
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis
Sigma-Aldrich
Copper(I) iodide, 98%