Merck
  • Structural investigations on sodium-lead borophosphate glasses doped with vanadyl ions.
Structural investigations on sodium-lead borophosphate glasses doped with vanadyl ions.
The journal of physical chemistry. A (2012-03-14)
K Srinivasulu, I Omkaram, H Obeid, A Suresh Kumar, J L Rao
ABSTRACT
Electron paramagnetic resonance (EPR), optical absorption, and FT-IR spectra of vanadyl ions in the sodium-lead borophosphate (Na(2)O-PbO-B(2)O(3)-P(2)O(5)) (SLBP) glass system have been studied. EPR spectra of all the glass samples exhibit resonance signals characteristic of VO(2+) ions. The spin Hamiltonian parameters g and A are found to be independent of the V(2)O(5) content and temperature. The values of the spin Hamiltonian parameters indicate that the VO(2+) ions in SLBP glasses are present in octahedral sites with tetragonal compression. The population difference between Zeeman levels (N) is calculated as a function of temperature for an SLBP glass sample containing 1.0 mol % VO(2+) ions. From the EPR data, the paramagnetic susceptibility (χ) is calculated at different temperatures, and the Curie constant (C) is calculated from the 1/χ versus T graph. The optical absorption spectra of the glass samples show two absorption bands, and they are attributed to V(3+) and V(4+) ions. The optical band gap energy (E(opt)) and Urbach energy (ΔE) are calculated from their ultraviolet absorption edges. It is observed that, as the vanadium ion concentration increases, E(opt) decreases and ΔE increases. The study of the IR absorption spectrum depicts the presence of BO(3), BO(4), PO(3), PO(4), and VO(5) structural units.
Sigma-Aldrich
Boric anhydride, puriss. p.a., ≥98% (T)
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Boric anhydride, 99.98% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Boric anhydride, 99.999% trace metals basis
Sigma-Aldrich
Boric anhydride, granulated, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lead(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Sodium oxide, 80%
Sigma-Aldrich
Lead(II) oxide, puriss. p.a., ≥99.0% (KT), yellow
Sigma-Aldrich
Lead(II) oxide, powder, <10 μm, ReagentPlus®, ≥99.9% trace metals basis
Sigma-Aldrich
Lead(II) oxide, ACS reagent, ≥99.0%