Merck
Biodegradation of 4-methylaniline in a sequencing batch reactor.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-03-02)
Iván Moreno-Andrade, Germán Buitrón
ABSTRACT
The biodegradation of the aromatic amine 4-methylaniline (4MA) using an aerobic sequencing batch reactor was evaluated. The specific degradation rate showed an exponential increase during the acclimation of the microorganisms reaching a maximal value of 34 mg 4MA/VSS/h. After the acclimation, the process showed a stable operation. A high similarity index was observed, indicating a low variation in the population diversity. During this period, the physicochemical parameters demonstrated a stable operation of the reactor.
Product Description
Supelco
N-Methylaniline, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylaniline, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methylaniline, ≥99%
Pricing and availability is not currently available.