Merck
Halothane changes the domain structure of a binary lipid membrane.
Langmuir : the ACS journal of surfaces and colloids (2012-02-23)
Michael Weinrich, Hirsh Nanda, David L Worcester, Charles F Majkrzak, Brian B Maranville, Sergey M Bezrukov
ABSTRACT
X-ray and neutron diffraction studies of a binary lipid membrane demonstrate that halothane at physiological concentrations produces a pronounced redistribution of lipids between domains of different lipid types identified by different lamellar d-spacings and isotope composition. In contrast, dichlorohexafluorocyclobutane (F6), a halogenated nonanesthetic, does not produce such significant effects. These findings demonstrate a specific effect of inhalational anesthetics on mixing phase equilibria of a lipid mixture.
Sigma-Aldrich
1,2-Didodecanoyl-sn-glycero-3-phosphocholine, ≥99%, synthetic