Merck
  • [The influence of viscosity of synthetic urine on the flow rate through catheters].
[The influence of viscosity of synthetic urine on the flow rate through catheters].
Der Urologe. Ausg. A (2012-02-22)
H Braunwarth, F H H Brill, D Hegeholz, P Hammerer
ABSTRACT
There are special requirements for draining a pouch, e.g. length, flexibility and ability to drain highly viscous urine. We checked in vitro the flow time of pure and synthetic urine thickened with hydroxyethylcellulose (0.125% and 0.250%, HEC) on selected catheters. We found for SpeediCath Compact Male (SCCM) as a CH12 catheter the shortest flow times for undiluted and urine thickened with 0.125% and for SafetyCath Plus for urine thickened with 0.250%. For catheters with CH14 we found significantly shorter flow times for all tested solutions. The flow time does not depend on amount and area of the catheter eyes; we found no difference in the flow time for 4-eye (MobiStom) and 2-eye catheters.
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~720,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~90,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~1,300,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mw ~380,000
Sigma-Aldrich
Hydroxyethyl-cellulose
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)