Merck
  • Protein dynamics in organic media at varying water activity studied by molecular dynamics simulation.
Protein dynamics in organic media at varying water activity studied by molecular dynamics simulation.
The journal of physical chemistry. B (2012-02-09)
Rasmus Wedberg, Jens Abildskov, Günther H Peters
ABSTRACT
In nonaqueous enzymology, control of enzyme hydration is commonly approached by fixing the thermodynamic water activity of the medium. In this work, we present a strategy for evaluating the water activity in molecular dynamics simulations of proteins in water/organic solvent mixtures. The method relies on determining the water content of the bulk phase and uses a combination of Kirkwood-Buff theory and free energy calculations to determine corresponding activity coefficients. We apply the method in a molecular dynamics study of Candida antarctica lipase B in pure water and the organic solvents methanol, tert-butyl alcohol, methyl tert-butyl ether, and hexane, each mixture at five different water activities. It is shown that similar water activity yields similar enzyme hydration in the different solvents. However, both solvent and water activity are shown to have profound effects on enzyme structure and flexibility.
Product Description
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Supelco
tert-Butanol, analytical standard
Sigma-Aldrich
tert-Butanol, puriss. p.a., ACS reagent, ≥99.7% (GC)
Sigma-Aldrich
tert-Butanol, ≥99% (GC)
Sigma-Aldrich
tert-Butanol, TEBOL® 99, ≥99.3%
Sigma-Aldrich
tert-Butanol, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butanol, suitable for HPLC, ≥99.5%