Merck
[Study on the chemical constituents of Phryma leptostachya].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-01-21)
Chun-Mei Chen, Hu-Cheng Zhu, De Zhao, Jun Deng
ABSTRACT
To study the chemical constituents of Phryma leptostachya. The chemical constituents were isolated and purified by silica gel column, recrystallization and Pre-RP-HPLC and their structwes were elucidated by spectroscopic methods. Five compounds were obtained and identified as ursolic acid (1), quercetin (2), isovanillic acid (3), p-hydroxybenzoic acid (4), adenine arabinoside (5). Compounds 2 - 5 are isolated from this genus for the first time.
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium 4-hydroxybenzoate, analytical standard