Merck
An in-the-ear platform for recording electroencephalogram.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2012-01-19)
D Looney, C Park, P Kidmose, M L Rank, M Ungstrup, K Rosenkranz, D P Mandic
ABSTRACT
We introduce a novel approach to brain monitoring based on electroencephalogram (EEG) recordings from within the ear canal. While existing clinical and wearable systems are limited in terms of portability and ease of use, the proposed in-the-ear (ITE) recording platform promises a number of advantages including ease of implementation, minimally intrusive electrodes and enhanced accuracy (fixed electrode positions). It thus facilitates a crucial step towards the design of brain computer interfaces that integrate naturally with daily life. The feasibility of the ITE concept is demonstrated with recordings made from electrodes embedded on an earplug which are benchmarked against conventional scalp electrodes for a classic EEG paradigm.
Sigma-Aldrich
Silver chloride, 99.999% trace metals basis
Sigma-Aldrich
Silver chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Silver chloride, ReagentPlus®, 99%