Merck
  • Bed nucleus of the stria terminalis and the cardiovascular responses to chemoreflex activation.
Bed nucleus of the stria terminalis and the cardiovascular responses to chemoreflex activation.
Autonomic neuroscience : basic & clinical (2011-12-27)
Erica M Granjeiro, Felipe V Gomes, Fernando H F Alves, Carlos C Crestani, Fernando M A Corrêa, Leonardo B M Resstel
ABSTRACT
Several studies from our group have indicated that the BNST play an important role in baroreflex modulation. However, the involvement of the BNST in the chemoreflex activity is unknown. Thus, in the present study, we investigated the effect of the local bed nucleus of stria terminalis (BNST) neurotransmission inhibition by bilateral microinjections of the non-selective synaptic blocker cobalt chloride (CoCl(2)) on the cardiovascular responses to chemoreflex activation in rats. For this purpose, chemoreflex was activated with KCN (i.v.) before and after microinjections of CoCl(2) into the BNST. Reversible BNST inactivation produced no significant changes in the magnitude and durations of both pressor and bradycardic responses to intravenous KCN infusion. These findings suggesting that BNST neurotransmission have not influence on both sympathoexcitatory and parasympathoexcitatory components of the peripheral chemoreflex activation.
Sigma-Aldrich
Potassium cyanide-15N, 98 atom % 15N
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
Potassium cyanide, technical, ≥96%
Sigma-Aldrich
Potassium cyanide, ACS reagent, ≥96.0%