Merck
  • Biosynthesis of gold nanoparticles by actinomycete Streptomyces viridogens strain HM10.
Biosynthesis of gold nanoparticles by actinomycete Streptomyces viridogens strain HM10.
Indian journal of biochemistry & biophysics (2011-12-15)
R Balagurunathan, M Radhakrishnan, R Babu Rajendran, D Velmurugan
ABSTRACT
Biosynthesis of gold nanoparticles by Streptomycetes from Himalayan Mountain was undertaken for the first time. Out of 10 actinomycete strains tested, four strains (D10, HM10, ANS2 and MSU) showed evidence for the intracellular biosynthesis of gold nanoparticles, among which the strain HM10 showed high potency. Presence of spherical and rod shaped gold nanoparticles in mycelium of the strain HM10 was determined by transmission electron microscopy (TEM) and X-ray diffraction analysis. The average particle size ranged from 18-20 nm. UV spectral analysis indicated that the reduction of chloroauric acid (HAuCl4) occurred within 24 h of reaction period. Further, the strain HM10 showed enhanced growth at 1 and 10 mM concentration of HAuCl4. The gold nanoparticles synthesized by the strain HM10 showed good antibacterial activity against S. aureus and E. coli in well-diffusion method. The potential actinomycete HM10 strain was phenotypically characterized and identified as Streptomyces viridogens (HM10). Thus, actinomycete strain HM10 reported in this study is a newly added source for the biosynthesis of gold nanoparticles.
Sigma-Aldrich
Sodium tetrachloroaurate(III) dihydrate, 99%
Sigma-Aldrich
Gold(III) chloride solution, 99.99% trace metals basis, 30 wt. % in dilute HCl
Sigma-Aldrich
Potassium gold(III) chloride, 98%
Sigma-Aldrich
Potassium gold(III) chloride, 99.995% trace metals basis