Merck
Quantification of sewer leakage by a continuous tracer method.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-11-08)
V Prigiobbe, M Giulianelli
ABSTRACT
Water authorities interested in the evaluation of the structural state of a sewer must quantify leakage to plan strategic intervention. However, the quantification of the exfiltration and the localisation of structural damage are challenging tasks that usually require expensive and time-consuming inspections. Herein, we report one of the first applications of the QUEST-C method to quantify the exfiltration in a continuously operating sewer by dosing two chemical tracers, sodium bromide (NaBr) and lithium chloride (LiCl). The method was applied at the catchment scale in a 14-year-old sewer in Rome, Italy. Preliminary laboratory tests, field measurements, and numerical simulations showed that reliable results require the QUEST-C method to be applied to sewers without lateral inflows, during periods of quasi-steady flow, and that the travel time of the NaBr tracer is minimised. Three sewer reaches were tested and the estimated exfiltration, as a fraction of the dry weather flow (DWF), increased from 0.128 in the agricultural area to 0.208 in the urban area. Although our estimates are at the lower end of the range given in the literature (0.01-0.56 DWF), the exfiltration was not negligible, and interventions should focus on the sewers in urban areas. This illustrates the capability of the QUEST-C method to guide strategic intervention at low cost and without an interruption of sewer operation. However, careful interpretation of the results is recommended for sewers with many lateral inflows, where leakage may be overestimated.
Product Description
Supelco
Density Standard 1251 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Sodium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium bromide, ReagentPlus®, ≥99%