Merck
  • Health based pasta: redefining the concept of the next generation convenience food.
Health based pasta: redefining the concept of the next generation convenience food.
Critical reviews in food science and nutrition (2011-10-14)
Murali Krishnan, P Prabhasankar
ABSTRACT
Pasta, a delicious meal favorite and the signature dish of many of the world's most famous chefs and a bonding comfort meal for millions all over the world, it has been recognized as an identifying ingredient of traditional healthy Mediterranean and Latin American meals. Pasta has come a long way from the days when it was erroneously considered by consumers to be a "fattening food." Today it is perceived as one of the "healthy options." In fact, because pasta is so supremely versatile as a base to a meal, it is easily possible to serve it in ways to satisfy both our notions of "healthy eating" and our appetites for interesting and tasty food. Pasta, being so popular as a delicious family meal favorite and equally relished all over the world, it deserves a lot than any other food to serve as an ideal functional food. Here we analyze various health ingredients that have been incorporated in pasta as disease/disorder curing agents and/or potent nutritional supplements and their effects on cooking and quality parameters of pasta as well as their various health benefits and therapeutic attributes.
Sigma-Aldrich
Amaranth, Dye content 85-95 %
Pricing and availability is not currently available.
Supelco
Amaranth, analytical standard
Pricing and availability is not currently available.