Merck
Magnetosomes on surface: an imaging study approach.
Scanning (2011-09-29)
Hubert Gojzewski, Marcin Makowski, Anezka Hashim, Peter Kopcansky, Zoltan Tomori, Milan Timko
ABSTRACT
In this study, we deposited isolated magnetosomes from magnetotactic bacteria Magnetospirillum strain AMB-1 onto solid surfaces using spin coating (SC) and drop coating (DC) techniques. Four imaging techniques have been used to visualize the sample structure: scanning and transmission electron microscopy (SEM, TEM), atomic and magnetic force microscopy (AFM, MFM). Additionally, dynamic light scattering was applied to measure the hydrodynamic radius of agglomerated/aggregated magnetosomes in a liquid environment. This manuscript discusses observed differences between structures obtained by two deposition techniques, i.e. possible interactions and factors responsible for magnetosomes' formation, their morphology on surfaces as a result of agglomeration and aggregation phenomena. Moreover, topography and homogeneity of obtained structures as well as thickness of protein-based membrane were also examined and described. Using high-resolution TEM, we analyzed the size of magnetic cores, their crystal structure and quality. We found that the SC technique provides a homogenous layer of magnetosomes and hydrophilization of silicon surfaces improves the deposition of magnetosomes. However, due to strong hydrogen interaction to the hydrophilic silicone surface, the organic membrane of magnetosomes is mostly flattened. As a matter of fact, the size distributions of magnetosomes deposited by SC and DC techniques (logarithmic-normal tendency) differ from the Feret diameter distribution (normal). Furthermore, our study confirms the good crystalline quality of magnetosomes' cores. It also shows that they are magnetic in the all their volume.
Product Description
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES sodium salt solution, 1M, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES sodium salt, Vetec, reagent grade, 96%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture