Merck
  • Separation and recycling of nanoparticles using cloud point extraction with non-ionic surfactant mixtures.
Separation and recycling of nanoparticles using cloud point extraction with non-ionic surfactant mixtures.
Journal of colloid and interface science (2011-08-27)
Muhammad Faizan Nazar, Syed Sakhawat Shah, Julian Eastoe, Asad Muhammad Khan, Afzal Shah
ABSTRACT
A viable cost-effective approach employing mixtures of non-ionic surfactants Triton X-114/Triton X-100 (TX-114/TX-100), and subsequent cloud point extraction (CPE), has been utilized to concentrate and recycle inorganic nanoparticles (NPs) in aqueous media. Gold Au- and palladium Pd-NPs have been pre-synthesized in aqueous phases and stabilized by sodium 2-mercaptoethanesulfonate (MES) ligands, then dispersed in aqueous non-ionic surfactant mixtures. Heating the NP-micellar systems induced cloud point phase separations, resulting in concentration of the NPs in lower phases after the transition. For the Au-NPs UV/vis absorption has been used to quantify the recovery and recycle efficiency after five repeated CPE cycles. Transmission electron microscopy (TEM) was used to investigate NP size, shape, and stability. The results showed that NPs are preserved after the recovery processes, but highlight a potential limitation, in that further particle growth can occur in the condensed phases.
Sigma-Aldrich
2-Mercaptoethanesulfonic acid solution, ampule, 3.0 M±0.1 M in H2O (T)
Sigma-Aldrich
Sodium 2-mercaptoethanesulfonate, BioXtra, ≥98.0% (RT)
Supelco
Sodium 2-mercaptoethanesulfonate, analytical standard, ≥98.0% (titration)