Merck
In vivo biocompatibility of three potential intraperitoneal implants.
Macromolecular bioscience (2011-08-09)
Sylvie Defrère, Mélanie Mestagdt, Raphaël Riva, Fabrice Krier, Anne Van Langendonckt, Pierre Drion, Christine Jérôme, Brigitte Evrard, Jean-Paul Dehoux, Jean-Michel Foidart, Jacques Donnez
ABSTRACT
The intraperitoneal biocompatibility of PDMS, polyHEMA and pEVA was investigated in rats, rabbits and rhesus monkeys. No inflammation was evidenced by hematological analyses and measurement of inflammatory markers throughout the experiment and by post-mortem examination of the pelvic cavity. After 3 or 6 months, histological analysis revealed fibrous tissue encapsulating PDMS and PEVA implants in all species and polyHEMA implants in rabbits and monkeys. Calcium deposits were observed inside polyHEMA implants. The intraperitoneal biocompatibility of all 3 polymers makes them suitable for the design of drug delivery systems, which may be of great interest for pathologies confined to the pelvic cavity.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor