Merck
  • Illness associated with exposure to methyl bromide-fumigated produce--California, 2010.
Illness associated with exposure to methyl bromide-fumigated produce--California, 2010.
MMWR. Morbidity and mortality weekly report (2011-07-15)
ABSTRACT
Methyl bromide (MeBr) is a toxic gas used to fumigate agricultural fields and some produce. The U.S. Department of Agriculture (USDA) requires MeBr fumigation of grapes imported from Chile to prevent invasion by the Chilean false red mite, Brevipalpus chilensis. In 2010, two workers were exposed intermittently to MeBr over several months as part of their job inspecting produce at a cold-storage facility in Carson, California. Both workers had disabling neurologic symptoms (e.g., ataxia, memory difficulties, and dizziness) and elevated serum bromide concentrations. An environmental investigation revealed the potential for MeBr to accumulate in enclosed areas during the transportation and storage of fumigated grapes. Some MeBr air concentrations measured at a single point in time exceeded current 8-hour exposure limits, suggesting that exposure in confined areas could result in poisoning. Possible measures for facilities managers to consider to reduce postfumigation MeBr exposures include 1) increased aeration time, 2) reduction of packaging that might absorb MeBr or limit aeration, and 3) changes in the stacking of pallets to improve air flow. Facilities should monitor air MeBr levels if they store MeBr-fumigated commodities in enclosed spaces entered by workers. Clinicians should consider occupational and environmental exposures in their differential diagnosis, and workers who might become exposed to fumigants should be informed of the health hazards related to these pesticides.
Sigma-Aldrich
Bromomethane, purum, ampule, ≥99.5% (GC)