Merck
  • Impact of agglomeration on the relaxometric properties of paramagnetic ultra-small gadolinium oxide nanoparticles.
Impact of agglomeration on the relaxometric properties of paramagnetic ultra-small gadolinium oxide nanoparticles.
Nanotechnology (2011-06-23)
Luc Faucher, Yves Gossuin, Aline Hocq, Marc-André Fortin
ABSTRACT
Ultra-small gadolinium oxide nanoparticles (US-Gd(2)O(3)) are used to provide 'positive' contrast effects in magnetic resonance imaging (MRI), and are being considered for molecular and cellular imaging applications. However, these nanoparticles can aggregate over time in aqueous medium, as well as when internalized into cells. This study is aimed at measuring in vitro, in aqueous medium, the impact of aggregation on the relaxometric properties of paramagnetic US-Gd(2)O(3) particles. First, the nanoparticle core size as well as aggregation behaviour was assessed by HRTEM. DLS (hydrodynamic diameter) was used to measure the hydrodynamic diameter of nanoparticles and nanoaggregates. The relaxometric properties were measured by NMRD profiling, as well as with (1)H NMR relaxometers. Then, the positive contrast enhancement effect was assessed by using magnetic resonance scanners (at 1.5 and 7 T). At every magnetic field, the longitudinal relaxivity (r(1)) decreased upon agglomeration, while remaining high enough to provide positive contrast. On the other hand, the transverse relaxivity (r(2)) slightly decreased at 0.47 and 1.41 T, but it was enhanced at higher fields (7 and 11.7 T) upon agglomeration. All NMRD profiles revealed a characteristic relaxivity peak in the range 60-100 MHz, suggesting the possibility to use US-Gd(2)O(3) as an efficient 'positive-T(1)' contrast agent at clinical magnetic fields (1-3 T), in spite of aggregation.
Product Description
Supelco
Diethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Diethylene glycol, analytical standard
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Gadolinium(III) oxide, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, nanopowder, <100 nm particle size (BET), 99.8% trace metals basis
Sigma-Aldrich
Gadolinium(III) oxide, ≥99.9%