Merck
  • [Malignant fascicular ventricular tachycardia degenerating into ventricular fibrillation in a patient with early repolarization syndrome].
[Malignant fascicular ventricular tachycardia degenerating into ventricular fibrillation in a patient with early repolarization syndrome].
Annales de cardiologie et d'angeiologie (2011-06-15)
Ad Kane, P Defaye, P Jacon, A Mbaye, J Machecourt
ABSTRACT
A 45-year-old man was hospitalized for syncope due to fascicular ventricular tachycardia degenerating into ventricular fibrillation (VF). The electrocardiogram showed an early repolarization syndrome. The arrhythmia was repetitive and disappeared after oral hydroquinidine. An implantable cardioverter-defibrillator (ICD) was implanted; subsequently, the patient was arrhythmia free at 9 months follow-up.
Sigma-Aldrich
Hydroquinine, 98%
Sigma-Aldrich
Hydroquinidine hydrochloride, 98%