Merck
  • Hydrothermal synthesis and properties of controlled α-Fe2O3 nanostructures in HEPES solution.
Hydrothermal synthesis and properties of controlled α-Fe2O3 nanostructures in HEPES solution.
Chemistry, an Asian journal (2011-06-11)
Hui Li, Zhong Lu, Qin Li, Man-Ho So, Chi-Ming Che, Rong Chen
ABSTRACT
A facile, template-free, and environmentally friendly hydrothermal strategy was explored for the controllable synthesis of α-Fe(2)O(3) nanostructures in HEPES solution (HEPES=2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulfonic acid). The effects of experimental parameters including HEPES/FeCl(3) molar ratio, pH value, reaction temperature, and reaction time on the formation of α-Fe(2)O(3) nanostructures have been investigated systematically. Based on the observations of the products, the function of HEPES in the reaction is discussed. The different α-Fe(2)O(3) nanostructures possess different optical, magnetic properties, and photocatalytic activities, depending on the shape and size of the sample. In addition, a novel and facile approach was developed for the synthesis of Au/α-Fe(2)O(3) and Ag/α-Fe(2)O(3) nanocomposites in HEPES buffer solution; this verified the dual function of HEPES both as reductant and stabilizer. This work provides a new strategy for the controllable synthesis of transition metal oxide nanostructures and metal-supported nanocomposites, and gives a strong evidence of the relationship between the property and morphology/size of nanomaterials.
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES sodium salt, Vetec, reagent grade, 96%
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture