Merck
  • Spectral studies on Ag8+ ions irradiated LAHCl·H2O and LAHBr·H2O single crystals.
Spectral studies on Ag8+ ions irradiated LAHCl·H2O and LAHBr·H2O single crystals.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-05-24)
K Sangeetha, R Ramesh Babu, K Ramamurthi, Jai Prakash, S A Khan
ABSTRACT
L-arginine hydrochloride monohydrate and L-arginine hydrobromide monohydrate single crystals are irradiated by 100 MeV Ag8+ swift heavy ions. The residual gases liberated from the irradiated samples are monitored as a function of ion fluence using quadrupole mass analyzer. The C2H3+, C2H2, N2, CO, HCl and CO2 are the dominant gases liberated. Fourier transform infrared spectra of irradiated crystals explain the breaking of bonds in a localized region of the crystals. The crystallinity of irradiated crystals is analyzed by powder X-ray diffractions.
Sigma-Aldrich
Hydrobromic acid, 48 wt. % in H2O, ≥99.99%
Sigma-Aldrich
Hydrogen bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrobromic acid, ACS reagent, 48%
Sigma-Aldrich
Hydrobromic acid, reagent grade, 48%