Merck
  • High-flux isobutanol production using engineered Escherichia coli: a bioreactor study with in situ product removal.
High-flux isobutanol production using engineered Escherichia coli: a bioreactor study with in situ product removal.
Applied microbiology and biotechnology (2011-05-07)
Antonino Baez, Kwang-Myung Cho, James C Liao
ABSTRACT
Promising approaches to produce higher alcohols, e.g., isobutanol, using Escherichia coli have been developed with successful results. Here, we translated the isobutanol process from shake flasks to a 1-L bioreactor in order to characterize three E. coli strains. With in situ isobutanol removal from the bioreactor using gas stripping, the engineered E. coli strain (JCL260) produced more than 50 g/L in 72 h. In addition, the isobutanol production by the parental strain (JCL16) and the high isobutanol-tolerant mutant (SA481) were compared with JCL260. Interestingly, we found that the isobutanol-tolerant strain in fact produced worse than either JCL16 or JCL260. This result suggests that in situ product removal can properly overcome isobutanol toxicity in E. coli cultures. The isobutanol productivity was approximately twofold and the titer was 9% higher than n-butanol produced by Clostridium in a similar integrated system.
Product Description
Supelco
2-Methyl-1-propanol, analytical standard
Sigma-Aldrich
2-Methyl-1-propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Methyl-1-propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2-Methyl-1-propanol, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, ACS reagent, ≥99.0%
Sigma-Aldrich
Isobutyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Isobutyl alcohol, natural, ≥99%, FCC, FG