Merck
  • The structure of human ubiquitin in 2-methyl-2,4-pentanediol: a new conformational switch.
The structure of human ubiquitin in 2-methyl-2,4-pentanediol: a new conformational switch.
Protein science : a publication of the Protein Society (2011-03-25)
Kuo Ying Huang, Gabriele A Amodeo, Liang Tong, Ann McDermott
ABSTRACT
A new crystal structure of human ubiquitin is reported at 1.8 Å resolution. Compared with the other known crystal structure or the solution NMR structure of monomeric human ubiquitin, this new structure is similar in its overall fold but differs with respect to the conformation of the backbone in a surface-exposed region. The conformation reported here resembles conformations previously seen in complex with deubiquinating enzymes, wherein the Asp52/Gly53 main chain and Glu24 side chain move. This movement exposes the backbone carbonyl of Asp52 to the exterior of the molecule, making it possible to engage in hydrogen-bond contacts with neighboring molecules, rather than in an internal hydrogen bond with the backbone of Glu24. This particular crystal form of ubiquitin has been used in a large number of solid state NMR studies. The structure described here elucidates the origin of many of the chemical shift differences comparing solution and solid state studies.
Sigma-Aldrich
Hexylene glycol, puriss., ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexylene glycol, BioUltra, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexylene glycol, BioXtra, ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hexylene glycol, 99%