Merck
  • Low-temperature methyl bromide fumigation of emerald ash borer (Coleoptera: Buprestidae) in ash logs.
Low-temperature methyl bromide fumigation of emerald ash borer (Coleoptera: Buprestidae) in ash logs.
Journal of economic entomology (2011-03-17)
Alan V Barak, Peggy Elder, Ivich Fraser
ABSTRACT
Ash (Fraxinus spp.) logs, infested with fully developed, cold-acclimated larval and prepupal emerald ash borer, Agrilus planipennis Fairmaire (Coleoptera: Buprestidae), were fumigated with methyl bromide (MeBr) at 4.4 and 10.0 degrees C for 24 h. Concentrations X time dosages of MeBr obtained were 1579 and 1273 g-h/m3 (24-h exposure) at 4.4 and 10.0 degrees C after applied doses of 112 and 96 g/m3, respectively. MeBr concentrations were simultaneously measured with a ContainIR infrared monitor and Fumiscope thermal conductivity meter calibrated for MeBr to measure the effect of CO2 on Fumiscope concentration readings compared with the infrared (IR) instrument. The presence of CO2 caused false high MeBr readings. With the thermal conductivity meter, CO2 measured 11.36 g/m3 MeBr per 1% CO2 in clean air, whereas the gas-specific infrared ContainIR instrument measured 9.55% CO2 as 4.2 g/m3 MeBr (0.44 g/m3 per 1% CO2). The IR instrument was 0.4% as sensitive to CO2 as the thermal conductivity meter. After aeration, fumigated and control logs were held for 8 wk to capture emerging beetles. No A. planipennis adults emerged from any of the fumigated logs, whereas 262 emerged from control logs (139 and 123/m2 at 4.4 and 10.0 degrees C, respectively). An effective fumigation dose and minimum periodic MeBr concentrations are proposed. The use of a CO2 scrubber in conjunction with nonspecific thermal conductivity instruments is necessary to more accurately measure MeBr concentrations.
Sigma-Aldrich
Bromomethane, purum, ampule, ≥99.5% (GC)