- A new peroxidase color reaction: oxidative coupling of 3-methyl-2-benzothiazolinone hydrazone (MBTH) with its formaldehyde azine. Application to glucose and choline oxidases.
A new peroxidase color reaction: oxidative coupling of 3-methyl-2-benzothiazolinone hydrazone (MBTH) with its formaldehyde azine. Application to glucose and choline oxidases.
Analytical biochemistry (1983-03-01)
D J Capaldi, K E Taylor
PMID6342472
ABSTRACT
Hydrogen peroxide in the presence of horseradish peroxidase effects the oxidative coupling of 3-methyl-2-benzothiazolinone hydrazone with its formaldehyde azine to form a tetraazapentamethine dye. The blue chromophore, when formed at pH 3.5 and quenched with acetone or 1 N hydrochloric acid, has an extinction coefficient of 69 +/- 2 or 55 +/- 2 mM-1 cm-1, respectively. This chromogen system has been adapted for enzymatic determinations of hydrogen peroxide and of glucose in the 10- to 45-nmol range and of choline in the 5- to 20-nmol range.