Merck
  • Fabrication of new solid state phosphate selective electrodes for environmental monitoring.
Fabrication of new solid state phosphate selective electrodes for environmental monitoring.
Talanta (2011-01-18)
Fikru Tafesse, Martin Enemchukwu
ABSTRACT
A highly selective and sensitive phosphate sensor has been fabricated by constructing a crystal disk consisting of variable mixtures of aluminium powder (Al), aluminium phosphate (AlPO(4)) and powdered copper (Cu). The membrane sensor exhibits linear potential response in the concentration range of 1.0×10(-1) to 1.0×10(-6) mol L(-1). The proposed sensor also exhibits a fast response time of <60s. Its detection limit is lower than 1.0×10(-6) mol L(-1). The electrode has a long lifetime and can be stored in air when not in use. The selectivity of the sensor with respect to other common ions is excellent.
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis