Merck
Aluminophosphates for CO₂ separation.
ChemSusChem (2011-01-13)
Qingling Liu, Ngo Chuen Ocean Cheung, Alfonso E Garcia-Bennett, Niklas Hedin
ABSTRACT
The pressure-swing adsorption method for carbon dioxide capture would ideally be facilitated by adsorbents with a high capacity and a high selectivity for CO₂. Several aluminophosphates with 8-ring window apertures (AlPO₄-17, AlPO₄-18, AlPO₄-53, and AlPO₄-25) were synthesized by hydrothermal crystallization, calcined, and their CO₂ uptake and CO₂/N₂ selectivity were studied. CO₂ and N₂ uptake was determined for pressures up to 101 kPa at 273 and 293 K. Langmuir and Toth adsorption models were used to describe the adsorption isotherms. The CO₂ and N₂ uptakes strongly indicated that the squeezed 8-ring windows of certain aluminophosphates can sieve CO₂ from a CO₂ and N₂ gas mixture. Both AlPO₄-53 and AlPO₄-25 exhibited a remarkably higher uptake of CO₂ compared to N₂. The hydrophilicity of the AlPO₄ materials was investigated by means of water adsorption, and the results showed that all of the tested aluminophosphates were less water sensitive than a benchmark zeolite (13X). In particular, AlPO₄-53 and AlPO₄-25 showed a very low degree of water uptake with up to 20-30 % relative humidity. Determination of cyclic adsorption and desorption confirmed the relatively hydrophobic nature of the aluminophosphates, which render them less energy costly for the regeneration of adsorbents.
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate, reagent grade